Potassium perruthenate(VII) ≥97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Potassium perruthenate
011877-06
011877-03
AA11877-06EA
888
USD
AA11877-06
AA11877-03
Potassium perruthenate(VII) ≥97%
Potassium perruthenate(VII)
Purity: 97%
CAS No.: 10378-50-4
Molecular Formula: KO4Ru
Formula Weight: 204.17
Physical Form: Crystalline
Appearance: Black
Melting Point: 440° dec.
UN No.: 1479
MDL No.: MFCD00061333
Formula:
C₃₂H₃₆N₂O₅ Melting Pt: 440 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00061333 CAS Number: 10378-50-4 EINECS: 233-835-6 UN: 1479 ADR: 5.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...