Molybdenum(VI) oxide ≥99.95% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

011837-A3 011837-36 011837-22
AA11837-A3EA 602 USD
AA11837-A3 AA11837-36 AA11837-22
Molybdenum(VI) oxide ≥99.95% (metals basis)
Formula: MoO₃
Boiling Pt: 1155 °C (subl.)
Melting Pt: 795 °C
Density: 4.692
Storage Temperature: Ambient
MDL Number: MFCD00003469
CAS Number: 1313-27-5
EINECS: 215-204-7
UN: 3288
ADR: 6.1,III
Merck Index: 14,6239

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR