Tungsten disulfide ≥99.8% (metals basis)
Supplier: Thermo Scientific Chemicals
Synonyms:
Tungsten sulfide, Tungsten sulphide, Tungsten (IV) sulfide, Tungsten (IV) sulphide
011829-18
011829-30
AA11829-18EA
88.9
USD
AA11829-18
AA11829-30
Tungsten disulfide ≥99.8% (metals basis)
Tungsten disulphide
MDL: MFCD00011467
Purity: 99.8%
CAS No.: 12138-09-9
Molecular Formula: WS2
Formula Weight: 247.98
Physical Form: Powder
Density: 7.5
Appearance: Black
Melting Point: 1250° dec.
MDL No.: MFCD00011467
Formula:
S₂W Melting Pt: 1250 °C (dec.) Density: 7,5 Storage Temperature: Ambient |
MDL Number:
MFCD00011467 CAS Number: 12138-09-9 EINECS: 235-243-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...