Ammonium iodide ≥99.0% ACS
Supplier: Thermo Scientific Chemicals
|
Warning
|
011668-22
AA11668-22EA
85.66
USD
AA11668-22
Ammonium iodide ≥99.0% ACS
Ammonium iodide
MDL: MFCD00011424
Freely soluble in water, alcohol, glycerol, methanol
Purity: 99.0% min
CAS No.: 12027-06-4
Molecular Formula: NH4I
Formula Weight: 144.94
Physical Form: Granular
Density: 2.514
Refractive Index: 1.7031
Sensitivity: Light and Air Sensitive and Hygroscopic
Appearance: White to pale yellow
Odor: Odorless
Melting Point: 551° subl.
Solubility: Freely soluble in water, alcohol, glycerol, methanol
Merck Reference: 14,529
MDL No.: MFCD00011424
Formula:
NH₄I Melting Pt: 551 °C (subl.) Density: 2.514 Storage Temperature: Ambient |
MDL Number:
MFCD00011424 CAS Number: 12027-06-4 EINECS: 234-717-7 Merck Index: 14,529 |
Specification Test Results
Insoluble matter | 0.005% |
Residue after ignition | 0.05% |
Chloride and bromide (as Cl) | 0.005% |
PO3 | 0.00001 |
SO3 | 0.05% |
Ba | 0.002% |
Heavy Metals (as Pb) | 0.001% |
Fe | 5 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...