Lead(II) perchlorate trihydrate 97.0-102.0% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
Titrant in complexometric titrations
Purity: 97% min
CAS No.: 13453-62-8
Molecular Formula: Cl2O8Pb.3H2O
Formula Weight: 460.15 (406.10anhy)
Physical Form: Crystalline
Density: 2.60
Appearance: White
Odor: Odorless
Melting Point: 100° dec.
Solubility: Soluble in water
UN No.: 1470
MDL No.: MFCD00149817
Titrant in complexometric titrations
Formula:
Cl₂O₈Pb·3H₂O Melting Pt: 100 °C (dec.) Density: 2.60 Storage Temperature: Ambient |
MDL Number:
MFCD00149817 CAS Number: 13453-62-8 EINECS: 237-125-7 UN: 1470 ADR: 5.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...