Copper(I) iodide 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Cuprous iodide
As a catalyst in organic synthesis, cathode ray tube coating, temperature indicator with HgI2
Purity: 98%
CAS No.: 7681-65-4
Molecular Formula: CuI
Formula Weight: 190.45
Physical Form: Powder
Density: 5.63
Refractive Index: 2.346
Sensitivity: Light, Air and Moisture Sensitive
Appearance: Pale brown
Odor: Weak
Melting Point: 605°
Boiling Point: ca 1290°
Solubility: Insoluble in water, dilute acids, alcohol. Decomposes by conc. acids. Soluble in NH3 solution, iodides, thiosulfates
Merck Reference: 14,2662
UN No.: 3077
MDL No.: MFCD00010978
As a catalyst in organic synthesis, cathode ray tube coating, temperature indicator with HgI2
Formula:
CuI Boiling Pt: ∼1290 °C Melting Pt: 605 °C Density: 5.63 Storage Temperature: Ambient |
MDL Number:
MFCD00010978 CAS Number: 7681-65-4 EINECS: 231-674-6 UN: 3077 ADR: 9,III Merck Index: 14,2662 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...