Zinc fluoride, anhydrous ≥99%

Supplier: Thermo Scientific Chemicals
Danger

011541-18 011541-A1 011541-30
AA11541-18EA 98.48 USD
AA11541-18 AA11541-A1 AA11541-30
Zinc fluoride, anhydrous ≥99%
Zinc fluoride

In phosphors, in ceramic glazes, in wood preserving, in electroplating, and in organic fluorination


Purity: 99% min CAS No.: 7783-49-5 Molecular Formula: ZnF2 Formula Weight: 103.37 Physical Form: Powder Density: 4.95 Appearance: White Melting Point: 872° Boiling Point: 1500° Solubility: Slightly soluble in water. Soluble in hot acids. Insoluble in alcohol Merck Reference: 14,10136 MDL No.: MFCD00011298


In phosphors, in ceramic glazes, in wood preserving, in electroplating, and in organic fluorination

Formula: F₂Zn
Boiling Pt: 1500 °C
Melting Pt: 872 °C
Density: 4.95
Storage Temperature: Ambient
MDL Number: MFCD00011298
CAS Number: 7783-49-5
EINECS: 232-001-9
UN: 3288
ADR: 6.1,III
Merck Index: 14,10136

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR