Lithium tetrafluoroborate 98%

Supplier: Thermo Scientific Chemicals
Danger

011528-18 011528-09
AA11528-18EA 174 USD
AA11528-18 AA11528-09
Lithium tetrafluoroborate 98%
Lithium tetrafluoroborate

MDL: MFCD00011087


Purity: 98% CAS No.: 14283-07-9 Molecular Formula: BF4Li Formula Weight: 93.75 Physical Form: Powder Sensitivity: Hygroscopic Appearance: White to off-white Odor: Odorless Melting Point: 293-300° dec. Merck Reference: 14,5543 UN No.: 3260 MDL No.: MFCD00011087

Formula: BF₄Li
MW: 93.74 g/mol
Melting Pt: 293…300 °C (dec.)
Density: 0.852 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011087
CAS Number: 14283-07-9
EINECS: 238-178-9
UN: 3260
ADR: 8 III
Merck Index: 14,5543

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR