Tin ≥99.998% (metals basis), foil, Puratronic®, Thickness 0.25 mm (0.01 in)

Supplier: Thermo Scientific Chemicals

011459-HP 011459-FI 011459-GH
AA11459-HPEA 700 USD
AA11459-HP AA11459-FI AA11459-GH
Tin ≥99.998% (metals basis), foil, Puratronic®, Thickness 0.25 mm (0.01 in)
Tin

MDL: MFCD00133862


Purity: 99.998% CAS No.: 7440-31-5 Physical Form: ≈4.6g/50x50mm Odor: Odorless MDL No.: MFCD00133862

Formula: Sn
Boiling Pt: 2270 °C (1013 hPa)
Melting Pt: 232 °C
Density: 7.31 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133862
CAS Number: 7440-31-5
EINECS: 231-141-8
Merck Index: 12,09587

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR