Tantalum ≥99.96% (metals basis), rods, Puratronic® (max. 0.005% Nb), Diameter 4.5 mm (0.18 in)

Supplier: Thermo Scientific Chemicals

011441-DM
AA11441-HREA 895.23 USD
AA11441-HR AA11441-DM
Tantalum ≥99.96% (metals basis), rods, Puratronic® (max. 0.005% Nb), Diameter 4.5 mm (0.18 in)
Tantalum

MDL: MFCD00011252


Purity: 99.96% CAS No.: 7440-25-7 Physical Form: ≈2.64g/cm Odor: Odorless MDL No.: MFCD00011252

Formula: Ta
MW: 180.95 g/mol
Boiling Pt: 5427 °C (1013 hPa)
Melting Pt: 2996 °C
Density: 16.69 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011252
CAS Number: 7440-25-7
EINECS: 231-135-5
Merck Index: 12,09223

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR