Ammonium cerium(IV) nitrate ≥99.5% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Scientific Chemicals
|
Danger
|
MDL: MFCD00151121
Fieser: 1,120 13,67 14,74 15,70 16,66 17,68 18,85 19,67 20,73 21,90
Purity: 99.5%
CAS No.: 16774-21-3
Molecular Formula: H8CeN8O18
Formula Weight: 548.23
Physical Form: Crystalline
Appearance: Orange
Odor: Pungent
Merck Reference: 14,1992
UN No.: 1477
MDL No.: MFCD00151121
Formula:
(NH₄)₂Ce(NO₃)₆ Melting Pt: 107…108 °C Density: 2.2 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00151121 CAS Number: 16774-21-3 EINECS: 240-827-6 UN: 1477 ADR: 5.1,II Merck Index: 14,1992 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...