Erbium(III) oxide ≥99.9% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Dierbium trioxide, Erbium oxide

011310-22 011310-A1 011310-14
AA11310-22EA 235 USD
AA11310-22 AA11310-A1 AA11310-14
Erbium(III) oxide ≥99.9% (REO, rare earth oxide basis), REacton®
Erbium (III) oxide

MDL: MFCD00010991


Purity: 99.9% CAS No.: 12061-16-4 Molecular Formula: Er2O3 Formula Weight: 382.56 Physical Form: Powder Density: 8.64 Melting Point: 2400° Merck Reference: 14,3646 MDL No.: MFCD00010991

Formula: Er₂O₃
Melting Pt: 2400 °C
Density: 8.64
Storage Temperature: Ambient
MDL Number: MFCD00010991
CAS Number: 12061-16-4
EINECS: 235-045-7
Merck Index: 14,3646

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR