Erbium(III) oxide ≥99.9% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Dierbium trioxide, Erbium oxide
011310-22
011310-A1
011310-14
AA11310-22EA
235
USD
AA11310-22
AA11310-A1
AA11310-14
Erbium(III) oxide ≥99.9% (REO, rare earth oxide basis), REacton®
Erbium (III) oxide
MDL: MFCD00010991
Purity: 99.9%
CAS No.: 12061-16-4
Molecular Formula: Er2O3
Formula Weight: 382.56
Physical Form: Powder
Density: 8.64
Melting Point: 2400°
Merck Reference: 14,3646
MDL No.: MFCD00010991
Formula:
Er₂O₃ Melting Pt: 2400 °C Density: 8.64 Storage Temperature: Ambient |
MDL Number:
MFCD00010991 CAS Number: 12061-16-4 EINECS: 235-045-7 Merck Index: 14,3646 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...