Holmium trichloride hexahydrate ≥99.9% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Scientific Chemicals
Warning

011277-22 011277-14
AA11277-22EA 407 USD
AA11277-22 AA11277-14
Holmium trichloride hexahydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Holmium trichloride hexahydrate

MDL: MFCD00149739


Purity: 99.9% CAS No.: 14914-84-2 Molecular Formula: Cl3Ho.6H2O Formula Weight: 379.38 (271.29anhy) Storage Temperature: 2°C to 8°C Physical Form: Crystalline Aggregates Sensitivity: Hygroscopic Odor: Odorless Melting Point: 718° Merck Reference: 14,4727 MDL No.: MFCD00149739

Formula: Cl₃Ho·6H₂O
Melting Pt: 718 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00149739
CAS Number: 14914-84-2
EINECS: 233-387-1
Merck Index: 14,4727

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR