Lanthanum(III) oxide ≥99.9% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Scientific Chemicals
|
Warning
|
011272-36
011272-22
AA11272-36EA
332
USD
AA11272-36
AA11272-22
Lanthanum(III) oxide ≥99.9% (REO, rare earth oxide basis), REacton®
Lanthanum (III) oxide
MDL: MFCD00011071
Purity: 99.9%
CAS No.: 1312-81-8
Molecular Formula: La2O3
Formula Weight: 325.82
Physical Form: Powder
Density: 6.51
Melting Point: 2307°
Boiling Point: 4200°
Merck Reference: 14,5363
MDL No.: MFCD00011071
Formula:
La₂O₃ Boiling Pt: 4200 °C Melting Pt: 2307 °C Density: 6.51 Storage Temperature: Ambient |
MDL Number:
MFCD00011071 CAS Number: 1312-81-8 EINECS: 215-200-5 Merck Index: 14,5363 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...