Lanthanum(III) chloride heptahydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Scientific Chemicals
|
Warning
|
MDL: MFCD00149756
Purity: 99.99%
CAS No.: 10025-84-0
Molecular Formula: LaCl3.7H2O
Formula Weight: 371.37 (245.27anhy)
Physical Form: Wet Crystalline Aggregates
Odor: Odorless
Merck Reference: 14,5363
MDL No.: MFCD00149756
Formula:
LaCl₃·7H₂O Boiling Pt: ∼860 °C (1013 hPa) Melting Pt: 91 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00149756 CAS Number: 10025-84-0 EINECS: 233-237-5 Merck Index: 14,5363 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...