Lutetium(III) oxide ≥99.9% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Lutetium oxide, Dilutetium trioxide

011255-14 011255-03 011255-06
AA11255-14EA 507.63 USD
AA11255-14 AA11255-03 AA11255-06
Lutetium(III) oxide ≥99.9% (REO, rare earth oxide basis), REacton®
Lutetium (III) oxide

MDL: MFCD00011100


Purity: 99.9% CAS No.: 12032-20-1 Molecular Formula: Lu2O3 Formula Weight: 397.94 Physical Form: Powder Density: 9.420 Melting Point: 2487° Merck Reference: 14,5615 MDL No.: MFCD00011100

Formula: Lu₂O₃
Melting Pt: 2487 °C
Density: 9.420
Storage Temperature: Ambient
MDL Number: MFCD00011100
CAS Number: 12032-20-1
EINECS: 234-764-3
Merck Index: 14,5615

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR