Neodymium(III) oxide ≥99.9% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Scientific Chemicals
Synonyms:
Dineodymium trioxide, Neodymium(III)-oxide, Neodymium oxide
011248-18
011248-A1
011248-30
AA11248-18EA
105
USD
AA11248-18
AA11248-A1
AA11248-30
Neodymium(III) oxide ≥99.9% (REO, rare earth oxide basis), REacton®
Neodymium (III) oxide
Powder. Purity based on rare earth contaminates.
Purity: 99.9%
CAS No.: 1313-97-9
Molecular Formula: Nd2O3
Formula Weight: 336.48
Physical Form: Powder
Density: 7.24
Melting Point: 2272°
Merck Reference: 14,6450
MDL No.: MFCD00011134
Formula:
Nd₂O₃ Boiling Pt: 3760 °C (1013 hPa) Melting Pt: 2272 °C Density: 7.24 Storage Temperature: Ambient |
MDL Number:
MFCD00011134 CAS Number: 1313-97-9 EINECS: 215-214-1 Merck Index: 14,6450 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...