Diboron trioxide, dehydrated (max. 0.02% H₂O) ≥99.999% (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Boron oxide, Boric anhydride
In the electronics industry as a liquid encapsulent for Czochralski-type growth of III-V semiconductor single crystals (e.g., GaAs, GaP, InP)
Purity: 99.999%
CAS No.: 1303-86-2
Molecular Formula: B2O3
Formula Weight: 69.62
Physical Form: Cylinder, 5.0cm dia, 55g/cylinder
Density: 2.46 +/- 0.01
Appearance: White
Odor: Odorless
Melting Point: ca 450°
Boiling Point: 1860°
Merck Reference: 14,1337
MDL No.: MFCD00011315
In the electronics industry as a liquid encapsulent for Czochralski-type growth of III-V semiconductor single crystals (e.g., GaAs, GaP, InP)
Formula:
B₂O₃ Boiling Pt: 1860 °C Melting Pt: ∼450 °C Density: 2.46 Storage Temperature: Ambient |
MDL Number:
MFCD00011315 CAS Number: 1303-86-2 EINECS: 215-125-8 REACH: 01-2119486655-24 Merck Index: 14,1337 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...