Germanium ≥99.9999% (metals basis), pieces, Puratronic® ≤ 2 cm (0.8 in)
Supplier: Thermo Scientific Chemicals
011144-06
011144-22
011144-14
AA11144-06EA
122
USD
AA11144-06
AA11144-22
AA11144-14
Germanium ≥99.9999% (metals basis), pieces, Puratronic® ≤ 2 cm (0.8 in)
Germanium
MDL: MFCD00085310
Purity: 99.9999+%
CAS No.: 7440-56-4
Physical Form: Electrical Resistivity:
Formula:
Ge Boiling Pt: 2830 °C (1013 hPa) Melting Pt: 937.2 °C Density: 5.323 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00085310 CAS Number: 7440-56-4 EINECS: 231-164-3 Merck Index: 12,04415 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...