Indium ≥99.9% (metals basis), tear drop shot 4 mm (0.2 in)

Supplier: Thermo Scientific Chemicals

011026-09 011026-18 011026-30
AA11026-09EA 79.37 USD
AA11026-09 AA11026-18 AA11026-30
Indium ≥99.9% (metals basis), tear drop shot 4 mm (0.2 in)
Indium

MDL: MFCD00134048


Purity: 99.9% CAS No.: 7440-74-6 Odor: Odorless MDL No.: MFCD00134048

Formula: In
MW: 114.82 g/mol
Boiling Pt: 2000
Melting Pt: 156
Density: 7.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134048
CAS Number: 7440-74-6
EINECS: 231-180-0
Merck Index: 13,04971

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR