Potassium fluoride ≥99.99% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

010980-04 010980-09 010980-18
AA10980-04EA 37.4 USD
AA10980-04 AA10980-09 AA10980-18
Potassium fluoride ≥99.99% (metals basis)
Potassium fluoride

MDL: MFCD00011398 Beilstein Registry No.: 3902818


Purity: 99.99% CAS No.: 7789-23-3 Molecular Formula: FK Formula Weight: 58.10 Physical Form: Crystalline Density: 2.48 Refractive Index: 1.363 Sensitivity: Hygroscopic Appearance: White Odor: Odorless Melting Point: 859° Boiling Point: 1505° Beilstein Reference: 3902818 Merck Reference: 14,7632 UN No.: 1812 MDL No.: MFCD00011398

Formula: KF
Boiling Pt: 1505 °C
Melting Pt: 859 °C
Density: 2.48
Storage Temperature: Ambient
MDL Number: MFCD00011398
CAS Number: 7789-23-3
EINECS: 232-151-5
UN: 1812
ADR: 6.1,III
Merck Index: 14,7632

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR