Gold ≥99.9985% (metals basis), wire, Premion®, Diameter 1.0 mm (0.04 in)

Supplier: Thermo Scientific Chemicals

010965-BW 010965-BP 010965-BS
AA10965-BWEA 2009 USD
AA10965-BW AA10965-BP AA10965-BS
Gold ≥99.9985% (metals basis), wire, Premion®, Diameter 1.0 mm (0.04 in)
Gold

MDL: MFCD00003436


Purity: 99.9985% CAS No.: 7440-57-5 Formula Weight: 196.97 Physical Form: ≈15.15g/m Odor: Odorless MDL No.: MFCD00003436

Formula: Au
MW: 196.97 g/mol
Boiling Pt: 2800
Melting Pt: 1063
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR