Cobalt ≥99.995% (metals basis), wire, Puratronic®, Diameter 2.0 mm (0.08 in)

Supplier: Thermo Scientific Chemicals
Danger

010945-BP 010945-BS 010945-BW
AA10945-BPEA 72.1 USD
AA10945-BP AA10945-BS AA10945-BW
Cobalt ≥99.995% (metals basis), wire, Puratronic®, Diameter 2.0 mm (0.08 in)
Cobalt

MDL: MFCD00010935


Purity: 99.995% CAS No.: 7440-48-4 Physical Form: ≈28.0g/m Odor: Odorless MDL No.: MFCD00010935

Formula: Co
Boiling Pt: 2870 °C (1013 hPa)
Melting Pt: 1495 °C
Density: 8.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010935
CAS Number: 7440-48-4
EINECS: 231-158-0
Merck Index: 12,02488

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR