Sodium nitrate ≥99.999% (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
|
Danger
|
MDL: MFCD00011119
Fieser: 18,336
Purity: 99.999%
CAS No.: 7631-99-4
Molecular Formula: NaNO3
Formula Weight: 84.99
Physical Form: Powder
Density: 2.26
Melting Point: 308°
Merck Reference: 14,8647
UN No.: 1498
MDL No.: MFCD00011119
Formula:
NaNO₃ Boiling Pt: 380 °C (1013 hPa) Melting Pt: 308 °C Density: 2.26 Storage Temperature: Ambient |
MDL Number:
MFCD00011119 CAS Number: 7631-99-4 EINECS: 231-554-3 UN: 1498 ADR: 5.1,III REACH: 01-2119488221-41 Merck Index: 14,8647 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...