Cobalt ≥99.995% (metals basis), rods, Puratronic®, Diameter 12 mm (0.47 in)

Supplier: Thermo Scientific Chemicals
Danger

010867-DJ 010867-HF
AA10867-DJEA 1330 USD
AA10867-DJ AA10867-HF
Cobalt ≥99.995% (metals basis), rods, Puratronic®, Diameter 12 mm (0.47 in)
Cobalt

Purity: 99.995% CAS No.: 7440-48-4 Physical Form: ≈10.1g/cm Odor: Odorless MDL No.: MFCD00010935

Formula: Co
Storage Temperature: Ambient
MDL Number: MFCD00010935
CAS Number: 7440-48-4
EINECS: 231-158-0
UN: 2809
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR