Silicon dioxide ≥99.999% (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Silicon(IV) oxide, Silicium dioxide, Silicium (IV) oxide
010856-22
010856-14
AA10856-22EA
550
USD
AA10856-22
AA10856-14
Silicon dioxide ≥99.999% (metals basis), Puratronic®
Silicium dioxide
MDL: MFCD00011232
Purity: 99.999%
CAS No.: 7631-86-9
Molecular Formula: SiO2
Formula Weight: 60.09
Physical Form: Powder
Appearance: White
Odor: Odorless
Melting Point: 1710°
Boiling Point: 2230°
Merck Reference: 14,8493
MDL No.: MFCD00011232
Formula:
SiO₂ Boiling Pt: 2230 °C Melting Pt: 1710 °C Density: 2.2 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011232 CAS Number: 7631-86-9 EINECS: 238-878-4 Merck Index: 14,8493 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...