Selenious acid ≥99.999% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Selenous acid

010855-14 010855-22
AA10855-14EA 323 USD
AA10855-14 AA10855-22
Selenious acid ≥99.999% (metals basis), Puratronic®
Selenious acid

MDL: MFCD00011331


Purity: 99.999% CAS No.: 7783-00-8 Molecular Formula: H2SeO3 Formula Weight: 128.97 Physical Form: Crystalline Aggregates Sensitivity: Hygroscopic Merck Reference: 14,8430 UN No.: 3290 MDL No.: MFCD00011331

Formula: H₂O₃Se
Boiling Pt: 315 °C (1013 hPa)
Melting Pt: 70 °C (dec.)
Density: 1.998 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011331
CAS Number: 7783-00-8
EINECS: 231-974-7
UN: 3290
ADR: 6.1,II
Merck Index: 14,8430

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR