Potassium dihydrogen phosphate ≥99.999% (metals basis), Puratronic® (Si ≤ 10 ppm)
Supplier: Thermo Scientific Chemicals
|
Warning
|
MDL: MFCD00011401
Notes: Loses H2O at 400°C, forming metaphosphate
Purity: 99.999%
CAS No.: 7778-77-0
Molecular Formula: H2KO4P
Formula Weight: 136.09
Physical Form: Crystalline powder
Density: 2.338
Appearance: White
Melting Point: 253°
Merck Reference: 14,7659
MDL No.: MFCD00011401
Formula:
KH₂PO₄ Boiling Pt: ∼100 °C (1013 hPa) Melting Pt: 253 °C Density: 2.338 Storage Temperature: Ambient |
MDL Number:
MFCD00011401 CAS Number: 7778-77-0 EINECS: 231-913-4 REACH: 01-2119490224-41 Merck Index: 14,7659 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...