Zinc 97,5% (metals basis), powder 4 - 7 µm

Supplier: Thermo Scientific Chemicals
Danger

010835-D9 010835-22 010835-A1
AA10835-D9EA 275 USD
AA10835-D9 AA10835-22 AA10835-A1
Zinc 97,5% (metals basis), powder 4 - 7 µm
Zinc

MDL: MFCD00011291


Purity: 97.5% CAS No.: 7440-66-6 Physical Form: Powder or flakes Density: 7.14 Appearance: blue-grey Odor: Odorless UN No.: 1436 MDL No.: MFCD00011291

Formula: Zn
MW: 65.39 g/mol
Boiling Pt: 908
Melting Pt: 420
Density: 7.14
Storage Temperature: Ambient
MDL Number: MFCD00011291
CAS Number: 7440-66-6
EINECS: 231-175-3
UN: 1436
ADR: 4.3 II
Merck Index: 12,10255

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR