Diammonium tetrachloropalladate ≥99.998% (metals basis) Pd 37%, Premion®
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Ammonium tetrachloropalladate(II, Ammonium tetrachloropalladate(II), Ammonium palladium (II) chloride, Ammonium tetrachloropalladate (II)
010824-06
010824-03
AA10824-06EA
612
USD
AA10824-06
AA10824-03
Diammonium tetrachloropalladate ≥99.998% (metals basis) Pd 37%, Premion®
Diammonium tetrachloropalladate
MDL: MFCD00010884
Purity: 99.998%
CAS No.: 13820-40-1
Molecular Formula: H8Cl4N2Pd
Formula Weight: 284.31
Physical Form: Powder
Density: 2.17
Appearance: Dark green to brown
Melting Point: dec.
MDL No.: MFCD00010884
Formula:
H₈Cl₄N₂Pd Melting Pt: Dec. Density: 2.17 Storage Temperature: Ambient |
MDL Number:
MFCD00010884 CAS Number: 13820-40-1 EINECS: 237-498-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...