Nickel (II) oxide ≥99.998% (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
|
Danger
|
MDL: MFCD00011145
Purity: 99.998%
CAS No.: 1313-99-1
Molecular Formula: NiO
Formula Weight: 74.71
Physical Form: Powder
Density: 6.67
Appearance: Dark green
Odor: Odorless
Melting Point: 1984°
Merck Reference: 14,6512
MDL No.: MFCD00011145
Formula:
NiO Melting Pt: 1984 °C Density: 6.67 Storage Temperature: Ambient |
MDL Number:
MFCD00011145 CAS Number: 1313-99-1 EINECS: 215-215-7 Merck Index: 14,6512 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...