Molybdenum(VI) oxide ≥99.998% (metals basis excluding W), Puratronic® (max. 300 ppm W)
Supplier: Thermo Scientific Chemicals
|
Danger
|
MDL: MFCD00003469
Purity: 99.998%
CAS No.: 1313-27-5
Molecular Formula: MoO3
Formula Weight: 143.94
Physical Form: Powder
Density: 4.692
Appearance: White to grey
Odor: Odorless
Melting Point: 795°
Boiling Point: 1155° subl.
Merck Reference: 14,6239
UN No.: 3288
MDL No.: MFCD00003469
Formula:
MoO₃ Boiling Pt: 1155 °C (subl.) Melting Pt: 795 °C Density: 4.692 Storage Temperature: Ambient |
MDL Number:
MFCD00003469 CAS Number: 1313-27-5 EINECS: 215-204-7 UN: 3288 ADR: 6.1,III Merck Index: 14,6239 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...