Sulfur ≥99.5%, powder, precipitated

Supplier: Thermo Scientific Chemicals
Danger

010785-36 010785-A3
AA10785-36EA 68.19 USD
AA10785-36 AA10785-A3
Sulfur ≥99.5%, powder, precipitated
Sulphur

MDL: MFCD00085316


Purity: 99.5% CAS No.: 7704-34-9 Odor: Weak UN No.: 1350 MDL No.: MFCD00085316

Formula: S
Boiling Pt: 445 °C (1013 hPa)
Melting Pt: 113…119 °C
Density: 2.36 g/cm³ (20 °C)
Flash Pt: 168 °C
Storage Temperature: Ambient
MDL Number: MFCD00085316
CAS Number: 7704-34-9
EINECS: 231-722-6
UN: 1350
ADR: 4.1,III
Merck Index: 13,09059

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR