Sulfur ≥99.999% (metals basis), pieces, Puratronic®
Supplier: Thermo Scientific Chemicals
|
Danger
|
010755-09
010755-18
010755-30
AA10755-09EA
45.48
USD
AA10755-09
AA10755-18
AA10755-30
Sulfur ≥99.999% (metals basis), pieces, Puratronic®
Sulphur
MDL: MFCD00085316
Purity: 99.999%
CAS No.: 7704-34-9
Physical Form: Random Sizes
Odor: Weak
UN No.: 1350
MDL No.: MFCD00085316
Formula:
S Boiling Pt: 445 °C (1013 hPa) Melting Pt: 113…119 °C Density: 2.36 g/cm³ (20 °C) Flash Pt: 168 °C Storage Temperature: Ambient |
MDL Number:
MFCD00085316 CAS Number: 7704-34-9 EINECS: 231-722-6 UN: 1350 ADR: 4.1,III Merck Index: 13,09059 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...