Lithium carbonate ≥99.998% (metals basis excluding Ca), Puratronic® (max. 0.002% Ca)

Supplier: Thermo Scientific Chemicals
Warning

010734-30 010734-A1 010734-18
AA10734-30EA 343 USD
AA10734-30 AA10734-A1 AA10734-18
Lithium carbonate ≥99.998% (metals basis excluding Ca), Puratronic® (max. 0.002% Ca)
Lithium carbonate

MDL: MFCD00011084


Purity: 99.998% CAS No.: 554-13-2 Molecular Formula: Li2CO3 Formula Weight: 73.90 Physical Form: Powder Density: 2.11 Appearance: White Odor: Odorless pH: 10-11.5 Melting Point: 723° Boiling Point: 1310° dec. Merck Reference: 14,5527 MDL No.: MFCD00011084

Formula: Li₂CO₃
MW: 73.89 g/mol
Boiling Pt: 1310 °C (dec.)
Melting Pt: 720
Density: 2.11
Storage Temperature: Ambient
MDL Number: MFCD00011084
CAS Number: 554-13-2
EINECS: 209-062-5
Merck Index: 14,5527

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR