Lead(II) nitrate ≥99.999% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals
Danger

010727-18 010727-30
AA10727-A1EA 1436.34 USD
AA10727-A1 AA10727-18 AA10727-30
Lead(II) nitrate ≥99.999% (metals basis), Puratronic®
Lead (II) nitrate

MDL: MFCD00011153


Purity: 99.999% CAS No.: 10099-74-8 Molecular Formula: Pb(NO3)2 Formula Weight: 331.20 Physical Form: Powder Density: 4.530 Melting Point: 470° dec. Merck Reference: 14,5414 UN No.: 1469 MDL No.: MFCD00011153

Formula: Pb(NO₃)₂
Melting Pt: 470 °C (dec.)
Density: 4.530
Storage Temperature: Ambient
MDL Number: MFCD00011153
CAS Number: 10099-74-8
EINECS: 233-245-9
UN: 1469
ADR: 5.1,II
Merck Index: 14,5414

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR