Lead(II) carbonate ≥99.999% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Lead carbonat

010721-14
AA10721-14EA 143 USD
AA10721-14
Lead(II) carbonate ≥99.999% (metals basis), Puratronic®
Lead (II) carbonate

MDL: MFCD00064631


Purity: 99.999% CAS No.: 598-63-0 Molecular Formula: PbCO3 Formula Weight: 267.20 Physical Form: Powder Odor: Odorless UN No.: 2291 MDL No.: MFCD00064631

Formula: PbCO₃
MW: 267.21 g/mol
Melting Pt: 315 °C
Density: 6.43 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064631
CAS Number: 598-63-0
EINECS: 209-943-4
UN: 2291
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR