Iron(III) oxide ≥99.995% (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Ferric oxide, Ferric Oxide, (iron(III) oxide), Diiron trioxide
010716-36
010716-22
010716-14
AA10716-36EA
2130
USD
AA10716-36
AA10716-22
AA10716-14
Iron(III) oxide ≥99.995% (metals basis), Puratronic®
Iron (III) oxide
MDL: MFCD00011008
Purity: 99.998%
CAS No.: 1309-37-1
Molecular Formula: Fe2O3
Formula Weight: 159.69
Physical Form: Powder
Density: 5.24
Appearance: Red-brown
Odor: Odorless
Melting Point: 1565°
Merck Reference: 14,4028
MDL No.: MFCD00011008
Formula:
Fe₂O₃ Melting Pt: 1565 °C Density: 5.24 Storage Temperature: Ambient |
MDL Number:
MFCD00011008 CAS Number: 1309-37-1 EINECS: 215-168-2 Merck Index: 14,4028 |
Specification Test Results
Total Metal Impurities | 0.002% max |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...