Rhenium ≥99.997% (metals basis), powder, Puratronic® -22 mesh

Supplier: Thermo Scientific Chemicals
Warning

010673-03 010673-06
AA10673-03EA 103 USD
AA10673-03 AA10673-06
Rhenium ≥99.997% (metals basis), powder, Puratronic® -22 mesh
Rhenium

MDL: MFCD00011195


Purity: 99.997% CAS No.: 7440-15-5 UN No.: 3089 MDL No.: MFCD00011195

Formula: Re
Storage Temperature: Ambient
MDL Number: MFCD00011195
CAS Number: 7440-15-5
EINECS: 231-124-5
UN: 3089
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR