Platinum ≥99.99% (metals basis), powder, Premion® -22 mesh

Supplier: Thermo Scientific Chemicals

010672-01 010672-03 010672-06
AA10672-01EA 99.2 USD
AA10672-01 AA10672-03 AA10672-06
Platinum ≥99.99% (metals basis), powder, Premion® -22 mesh
Platinum

MDL: MFCD00011179


Purity: 99.99% CAS No.: 7440-06-4 Appearance: Silver-white Odor: Odorless MDL No.: MFCD00011179

Formula: Pt
Boiling Pt: 3827 °C (1013 hPa)
Melting Pt: 1769 °C
Density: 21.447 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS Number: 7440-06-4
EINECS: 231-116-1
Merck Index: 12,07689

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR