Barium nitrate ≥99.999% (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
|
Warning
|
010648-36
010648-22
010648-06
010648-14
AA10648-36EA
658
USD
AA10648-36
AA10648-22
AA10648-06
AA10648-14
Barium nitrate ≥99.999% (metals basis), Puratronic®
Barium nitrate
MDL: MFCD00003442
Purity: 99.999%
CAS No.: 10022-31-8
Molecular Formula: BaN2O6
Formula Weight: 261.35
Physical Form: Crystalline
Density: 3.24
Appearance: White
Melting Point: 592° dec.
Merck Reference: 14,983
UN No.: 1446
MDL No.: MFCD00003442
Formula:
Ba(NO₃)₂ Melting Pt: 592 °C (dec.) Density: 3.24 Storage Temperature: Ambient |
MDL Number:
MFCD00003442 CAS Number: 10022-31-8 EINECS: 233-020-5 UN: 1446 ADR: 5.1,II Merck Index: 14,983 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...