Antimony (V) oxide ≥99.998% (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Antimony pentoxide, Diantimony pentoxide
010643-22
010643-06
010643-14
AA10643-22EA
583
USD
AA10643-22
AA10643-06
AA10643-14
Antimony (V) oxide ≥99.998% (metals basis), Puratronic®
Antimony (V) oxide
MDL: MFCD00011216
Slightly soluble in water. Slowly dissolves in warm HCl and warm KOH. Insoluble in HNO3
Purity: 99.998%
CAS No.: 1314-60-9
Molecular Formula: Sb2O5
Formula Weight: 323.50
Physical Form: Powder
Density: 3.78
Melting Point: 380° dec.
Solubility: Slightly soluble in water. Slowly dissolves in warm HCl and warm KOH. Insoluble in HNO3
Merck Reference: 14,698
UN No.: 1549
MDL No.: MFCD00011216
Formula:
O₅Sb₂ Melting Pt: 380 °C (dec.) Density: 3.78 Storage Temperature: Ambient |
MDL Number:
MFCD00011216 CAS Number: 1314-60-9 EINECS: 215-237-7 UN: 1549 ADR: 6.1,III Merck Index: 14,698 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...