Aluminium nitrate hydrate ≥99.999% (metals basis excluding Hg), Puratronic®

Supplier: Thermo Scientific Chemicals
Danger

010626-36 010626-06 010626-22 010626-14
AA10626-36EA 827 USD
AA10626-36 AA10626-06 AA10626-22 AA10626-14
Aluminium nitrate hydrate ≥99.999% (metals basis excluding Hg), Puratronic®
Aluminium nitrate hydrate

MDL: MFCD00003419


Purity: 99.999% CAS No.: 25838-59-9 Molecular Formula: Al(NO3)3.xH2O (x~9) Formula Weight: 212.99(anhy) Physical Form: Crystalline Sensitivity: Hygroscopic Appearance: White Odor: Odorless pH: 2.5-3.5 Melting Point: 73.5° Merck Reference: 14,352 UN No.: 1438 MDL No.: MFCD00003419

Formula: AlN₃O₉·H₂O
MW: 231.01 g/mol
Melting Pt: 73.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00003419
CAS Number: 25838-59-9
EINECS: 236-751-8
UN: 1438
ADR: 5.1 III
Merck Index: 14,352

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR