Indium ≥99.9999% (metals basis), shot, Puratronic® ≤ 5 mm (0.2 in)

Supplier: Thermo Scientific Chemicals

010615-09 010615-30 010615-18
AA10615-09EA 139 USD
AA10615-09 AA10615-30 AA10615-18
Indium ≥99.9999% (metals basis), shot, Puratronic® ≤ 5 mm (0.2 in)
Indium

MDL: MFCD00134048


Purity: 99.9999% CAS No.: 7440-74-6 Odor: Odorless MDL No.: MFCD00134048

Formula: In
MW: 114.82 g/mol
Boiling Pt: 2000
Melting Pt: 156
Density: 7.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134048
CAS Number: 7440-74-6
EINECS: 231-180-0
Merck Index: 13,04971

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR