Gold ≥99.99%, powder, Premion®

Supplier: Thermo Scientific Chemicals

010612-03 010612-06
AA10612-03EA 287 USD
AA10612-03 AA10612-06
Gold ≥99.99%, powder, Premion®
Gold

MDL: MFCD00003436


Purity: 99.99% CAS No.: 7440-57-5 Appearance: Gold MDL No.: MFCD00003436

Formula: Au
MW: 196.97 g/mol
Boiling Pt: 2800
Melting Pt: 1063
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR