Selenium ≥99.5% (metals basis), powder -325 mesh

Supplier: Thermo Scientific Chemicals
Danger

010601-30 010601-06 010601-18
AA10601-30EA 510 USD
AA10601-30 AA10601-06 AA10601-18
Selenium ≥99.5% (metals basis), powder -325 mesh
Selenium

MDL: MFCD00134090


Purity: 99.5% CAS No.: 7782-49-2 UN No.: 3077 MDL No.: MFCD00134090

Formula: Se
MW: 78.96 g/mol
Boiling Pt: 685
Melting Pt: 217
Density: 4.28 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134090
CAS Number: 7782-49-2
EINECS: 231-957-4
UN: 3288
ADR: 6.1 III
Merck Index: 12,08572

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR