Selenium ≥99.5% (metals basis), powder -325 mesh
Supplier: Thermo Scientific Chemicals
|
Danger
|
010601-30
010601-06
010601-18
AA10601-30EA
510
USD
AA10601-30
AA10601-06
AA10601-18
Selenium ≥99.5% (metals basis), powder -325 mesh
Selenium
MDL: MFCD00134090
Purity: 99.5%
CAS No.: 7782-49-2
UN No.: 3077
MDL No.: MFCD00134090
Formula:
Se MW: 78.96 g/mol Boiling Pt: 685 Melting Pt: 217 Density: 4.28 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00134090 CAS Number: 7782-49-2 EINECS: 231-957-4 UN: 3288 ADR: 6.1 III Merck Index: 12,08572 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...