Ruthenium(III) acetylacetonate Ru ≥ 24%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ruthenium (III) 2,4-pentanedionate, Tris(2,4-pentanedionato-O,O')ruthenium (III), Ruthenium (III) tris(2,4-pentanedionato-O,O'), Tris(2,4-pentanedionato)ruthenium(III)

010568-06 010568-03
AA10568-06EA 414 USD
AA10568-06 AA10568-03
Ruthenium(III) acetylacetonate Ru ≥ 24%
Ruthenium (III) acetylacetonate

Hydrogenation


CAS No.: 14284-93-6 Molecular Formula: C15H21O6Ru Formula Weight: 398.40 Physical Form: Crystalline Appearance: Red Melting Point: 260° dec. MDL No.: MFCD00000030


Hydrogenation

Formula: C₁₅H₂₁O₆Ru
Melting Pt: 260 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00000030
CAS Number: 14284-93-6
EINECS: 238-193-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR