Rubidium carbonate ≥99.8% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

010562-09 010562-18
AA10562-09EA 70 USD
AA10562-09 AA10562-18
Rubidium carbonate ≥99.8% (metals basis)
Rubidium carbonate

MDL: MFCD00011188


Purity: 99.8% CAS No.: 584-09-8 Molecular Formula: CO3Rb2 Formula Weight: 230.95 Physical Form: -20 Mesh Powder Sensitivity: Hygroscopic Appearance: White Odor: Odorless Melting Point: 837° MDL No.: MFCD00011188

Formula: CO₃Rb₂
Melting Pt: 837 °C
Density: 2,43 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011188
CAS Number: 584-09-8
EINECS: 209-530-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR